4-Bromo-o-anisidine 97%

Supplier: Ambeed
Warning

A126730-10G A126730-25G A126730-500G A126730-5G A126730-100G
77108-908EA 13.39 USD
77108-908 77108-910 77108-912 77108-914 77592-458
4-Bromo-o-anisidine 97%
4-Bromo-o-anisidine

Precaución: for Research Use Only

Formula: C₇H₈BrNO
MDL Number: MFCD01204266
CAS Number: 59557-91-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR