2-Cyclopentylglycine 95%

Supplier: Ambeed
Warning

A126432-10G A126432-1G A126432-250MG A126432-5G
77108-874EA 203.94 USD
77108-874 77108-876 77108-878 77108-880
2-Cyclopentylglycine 95%
2-Cyclopentylglycine

Precaución: for Research Use Only

Formula: C₇H₁₃NO₂
MDL Number: MFCD09264351
CAS Number: 933-95-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR