1,4-Dibromo-2,5-dichlorobenzene 95%

Supplier: Ambeed
Warning

A126296-1G A126296-250MG A126296-5G
77108-868EA 36.04 USD
77108-868 77108-870 77108-872
1,4-Dibromo-2,5-dichlorobenzene 95%
1,4-Dibromo-2,5-dichlorobenzene

Precaución: for Research Use Only

Formula: C₆H₂Br₂Cl₂
MW: 304.79 g/mol
MDL Number: MFCD00182443
CAS Number: 4571-24-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR