Methyl-2-amino-5-bromonicotinate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 2-amino-5-bromopyridine-3-carboxylate
A125931-100G
A125931-10G
A125931-1G
A125931-25G
A125931-500G
A125931-5G
77108-846EA
151.71
USD
77108-846
77108-848
77108-850
77108-852
77108-854
77108-856
Methyl-2-amino-5-bromonicotinate 95%
Methyl-2-amino-5-bromonicotinate
Precaución: for Research Use Only
Formula:
C₇H₇BrN₂O₂ MW: 231.05 g/mol |
MDL Number:
MFCD04117787 CAS Number: 50735-34-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...