Sodium 3-(3,4-dihydroxyphenyl)-2-hydroxypropanoate 98+%

Supplier: Ambeed
Warning

A125917-100MG A125917-10MG A125917-1MG A125917-250MG A125917-50MG A125917-1G
77108-836EA 29.86 USD
77108-836 77108-838 77108-840 77108-842 77108-844 77685-100
Sodium 3-(3,4-dihydroxyphenyl)-2-hydroxypropanoate 98+%
Sodium 3-(3,4-dihydroxyphenyl)-2-hydroxypropanoate

Precaución: for Research Use Only

Formula: C₉H₉NaO₅
MDL Number: MFCD09037393
CAS Number: 67920-52-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR