Methyl-5-cyano-2-hydroxybenzenecarboxylate 97%

Supplier: Ambeed
Warning

A125887-100MG A125887-1G
77108-832EA 11.34 USD
77108-832 77108-834
Methyl-5-cyano-2-hydroxybenzenecarboxylate 97%
Methyl-5-cyano-2-hydroxybenzenecarboxylate

Precaución: for Research Use Only

Formula: C₉H₇NO₃
MDL Number: MFCD00800949
CAS Number: 84437-12-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR