3-Nitro-1H-pyrazole 98%

Supplier: Ambeed
Warning

A125626-100G A125626-10G A125626-500G A125626-5G A125626-25G
77108-818EA 47.75 USD
77108-818 77108-820 77108-822 77108-824 77312-852
3-Nitro-1H-pyrazole 98%
3-Nitro-1H-pyrazole

Precaución: for Research Use Only

Formula: C₃H₃N₃O₂
MW: 113.08 g/mol
MDL Number: MFCD00238787
CAS Number: 26621-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR