(Iodomethyl)triphenylphosphonium iodide 97%

Supplier: Ambeed
Warning

A125527-1G A125527-25G A125527-5G A125527-100G
77108-812EA 6.37 USD
77108-812 77108-814 77108-816 77566-416
(Iodomethyl)triphenylphosphonium iodide 97%
(Iodomethyl)triphenylphosphonium iodide

Precaución: For research use only.

Formula: C₁₉H₁₇I₂P
MW: 530.12 g/mol
MDL Number: MFCD09025335
CAS Number: 3020-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR