6-Aminonicotinamide 97%

Supplier: Ambeed
Warning

Synonyms: 6-Aminopyridine-3-carboxamide

A124658-10G A124658-1G A124658-250MG A124658-5G A124658-25G
77108-766EA 204.76 USD
77108-766 77108-768 77108-770 77108-772 77685-091
6-Aminonicotinamide 97%
6-Aminonicotinamide

Precaución: for Research Use Only

Formula: C₆H₇N₃O
MW: 137.14 g/mol
MDL Number: MFCD00006327
CAS Number: 329-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR