(R)-2-Amino-4,4-dimethylpentanoic acid 97%

Supplier: Ambeed
Warning

Synonyms: H-D-4-Me-Leu-OH, H-β-tBu-D-Ala-OH

A124532-1G A124532-250MG
77108-756EA 157.02 USD
77108-756 77108-758
(R)-2-Amino-4,4-dimethylpentanoic acid 97%
(R)-2-Amino-4,4-dimethylpentanoic acid

Precaución: for Research Use Only

Formula: C₇H₁₅NO₂
MW: 145.202 g/mol
MDL Number: MFCD00038404
CAS Number: 88319-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR