PF-670462 98%

Supplier: Ambeed
Warning

Synonyms: 4-(1-Cyclohexyl-4-(4-fluorophenyl)-1H-imidazol-5-yl)pyrimidin-2-amine dihydrochloride, 4-[1-Cyclohexyl-4-(4-fluorophenyl)-1H-imidazol-5-yl]-2-pyrimidinamine dihydrochloride, PF-670462 2HCl

A124173-10MG A124173-1MG A124173-50MG A124173-5MG
77108-744EA 62.6 USD
77108-744 77108-746 77108-748 77108-750
PF-670462 98%
PF-670462

Precaución: for Research Use Only

Formula: C₁₉H₂₀FN₅.2HCl
MW: 410.32 g/mol
MDL Number: MFCD12828759
CAS Number: 950912-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR