3-(4-Bromophenyl)butanoic acid 95%

Supplier: Ambeed
Warning

A123965-100MG A123965-1G A123965-250MG A123965-5G
77108-736EA 92.3 USD
77108-736 77108-738 77108-740 77108-742
3-(4-Bromophenyl)butanoic acid 95%
3-(4-Bromophenyl)butanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₂
MW: 243.09 g/mol
MDL Number: MFCD08282290
CAS Number: 53086-46-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR