(±)-Ethyl mandelate 95%

Supplier: Ambeed
Warning

A123892-100G A123892-10G A123892-25G A123892-500G A123892-5G
77108-718EA 98.88 USD
77108-718 77108-720 77108-722 77108-724 77108-726
(±)-Ethyl mandelate 95%
(±)-Ethyl mandelate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MDL Number: MFCD00004494
CAS Number: 774-40-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR