3-Bromo-N,N-dimethylpyridin-4-amine 95%

Supplier: Ambeed
Warning

A123782-250MG A123782-1G
77108-716EA 38.12 USD
77108-716 77685-086
3-Bromo-N,N-dimethylpyridin-4-amine 95%
3-Bromo-N,N-dimethylpyridin-4-amine

Precaución: for Research Use Only

Formula: C₇H₉BrN₂
MDL Number: MFCD09743635
CAS Number: 84539-35-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR