Ethylvanillin 98%

Supplier: Ambeed
Warning

Synonyms: 3-Ethoxy-4-hydroxybenzaldehyde, 3-(Ethoxy-d5)-4-hydroxybenzaldehyde

A123299-100G A123299-10G A123299-25G A123299-500G A123299-1000G
77108-692EA 19.1 USD
77108-692 77108-694 77108-696 77108-698 77685-085
Ethylvanillin 98%
Ethylvanillin

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
MDL Number: MFCD00006944
CAS Number: 121-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR