4,4'-Oxybis(butan-1-ol) 95%

Supplier: Ambeed
Warning

Synonyms: 4,4'-Oxybis(butan-1-ol)

A123229-100MG A123229-1G A123229-250MG
77108-686EA 50.93 USD
77108-686 77108-688 77108-690
4,4'-Oxybis(butan-1-ol) 95%
4,4'-Oxybis(butan-1-ol)

Precaución: for Research Use Only

Formula: C₈H₁₈O₃
MW: 162.23 g/mol
MDL Number: MFCD19982829
CAS Number: 3403-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR