6-Bromochroman 98%

Supplier: Ambeed
Warning

A122679-100MG A122679-1G A122679-250MG A122679-5G
77108-652EA 9.27 USD
77108-652 77108-654 77108-656 77108-658
6-Bromochroman 98%
6-Bromochroman

Precaución: for Research Use Only

Formula: C₉H₉BrO
MDL Number: MFCD10698725
CAS Number: 3875-78-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR