4-Nitrobenzyl alcohol 97%
Supplier: Ambeed
|
Warning
|
A122064-1000G
A122064-100G
A122064-10G
A122064-25G
A122064-500G
77108-642EA
48.41
USD
77108-642
77108-644
77108-646
77108-648
77108-650
4-Nitrobenzyl alcohol 97%
4-Nitrobenzyl alcohol
Precaución: for Research Use Only
Formula:
C₇H₇NO₃ |
MDL Number:
MFCD00007376 CAS Number: 619-73-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...