6-Methylpyridine-2-boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Borono-6-methylpyridine, (6-Methylpyridin-2-yl)boronic acid

A122047-100MG A122047-1G A122047-250MG
77108-636EA 58.35 USD
77108-636 77108-638 77108-640
6-Methylpyridine-2-boronic acid 97%
6-Methylpyridine-2-boronic acid

Precaución: for Research Use Only

Formula: C₆H₈BNO₂
MW: 136.944 g/mol
MDL Number: MFCD03093331
CAS Number: 372963-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR