N-Boc-L-tryptophan benzyl ester 97%

Supplier: Ambeed
Warning

Synonyms: Boc-Trp-OBzl

A122041-100G A122041-1G A122041-250MG A122041-5G
77108-628EA 542.81 USD
77108-628 77108-630 77108-632 77108-634
N-Boc-L-tryptophan benzyl ester 97%
N-Boc-L-tryptophan benzyl ester

Precaución: for Research Use Only

Formula: C₂₃H₂₆N₂O₄
MW: 394.471 g/mol
MDL Number: MFCD00190837
CAS Number: 57229-67-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR