6-Hydroxy-5-nitronicotinic acid 98%

Supplier: Ambeed
Warning

A121951-100G A121951-10G A121951-1G A121951-25G A121951-5G
77108-618EA 211.12 USD
77108-618 77108-620 77108-622 77108-624 77108-626
6-Hydroxy-5-nitronicotinic acid 98%
6-Hydroxy-5-nitronicotinic acid

Precaución: for Research Use Only

Formula: C₆H₄N₂O₅
MW: 184.1100 g/mol
MDL Number: MFCD02180840
CAS Number: 6635-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR