4-Cyanophenylacetonitrile 95%

Supplier: Ambeed
Warning

A121766-1G A121766-250MG A121766-25G A121766-5G
77108-610EA 7.21 USD
77108-610 77108-612 77108-614 77108-616
4-Cyanophenylacetonitrile 95%
4-Cyanophenylacetonitrile

Precaución: for Research Use Only

Formula: C₉H₆N₂
MDL Number: MFCD00060305
CAS Number: 876-31-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR