Ethyl-2-(5-bromopyrimidin-2-yl)acetate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-(5-bromopyrimidin-2-yl)acetate

A121383-1G A121383-5G
77108-602EA 43.5 USD
77108-602 77571-272
Ethyl-2-(5-bromopyrimidin-2-yl)acetate 97%
Ethyl-2-(5-bromopyrimidin-2-yl)acetate

Precaución: for Research Use Only

Formula: C₈H₉BrN₂O₂
MW: 245.0733 g/mol
MDL Number: MFCD09909703
CAS Number: 1134327-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR