1-Bromo-4-chloro-3-fluorobenzene 98%

Supplier: Ambeed
Warning

A120935-100G A120935-10G A120935-25G A120935-500G A120935-5G
77108-592EA 42.23 USD
77108-592 77108-594 77108-596 77108-598 77108-600
1-Bromo-4-chloro-3-fluorobenzene 98%
1-Bromo-4-chloro-3-fluorobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrClF
MDL Number: MFCD00042519
CAS Number: 60811-18-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR