(R)-tert-Butyl (1-([1,1'-biphenyl]-4-yl)-3-hydroxypropan-2-yl)carbamate 95%

Supplier: Ambeed
Warning

A120802-10G A120802-1G A120802-25G
77108-584EA 35.01 USD
77108-584 77108-586 77108-588 77108-590
(R)-tert-Butyl (1-([1,1'-biphenyl]-4-yl)-3-hydroxypropan-2-yl)carbamate 95%
(R)-tert-Butyl (1-([1,1'-biphenyl]-4-yl)-3-hydroxypropan-2-yl)carbamate

Precaución: for Research Use Only

Formula: C₂₀H₂₅NO₃
MDL Number: MFCD28359221
CAS Number: 1426129-50-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR