2-Bromo-4-fluorobenzonitrile 97%

Supplier: Ambeed
Danger

A120631-10G A120631-25G A120631-500G A120631-5G
77108-576EA 9.26 USD
77108-576 77108-578 77108-580 77108-582
2-Bromo-4-fluorobenzonitrile 97%
2-Bromo-4-fluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃BrFN
MDL Number: MFCD00672924
CAS Number: 36282-26-5
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR