6-Aminonicotinic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Aminopyridine-3-carboxylic acid, 6-Amino-3-pyridinecarboxylic acid
A120511-100G
A120511-10G
A120511-25G
A120511-5G
77108-550EA
78.51
USD
77108-550
77108-552
77108-554
77108-556
6-Aminonicotinic acid 98%
6-Aminonicotinic acid
Precaución: for Research Use Only
Formula:
C₆H₆N₂O₂ MW: 138.13 g/mol |
MDL Number:
MFCD00006326 CAS Number: 3167-49-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...