Fmoc-Hyp-OH 98%

Supplier: Ambeed
Warning

Synonyms: N-Fmoc-trans-4-hydroxy-L-proline

A120480-100G A120480-10G A120480-1G A120480-25G A120480-500G A120480-5G
77108-538EA 80.34 USD
77108-538 77108-540 77108-542 77108-544 77108-546 77108-548
Fmoc-Hyp-OH 98%
Fmoc-Hyp-OH

Precaución: for Research Use Only

Formula: C₂₀H₁₉NO₅
MW: 353.374 g/mol
MDL Number: MFCD00151929
CAS Number: 88050-17-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR