Tetramethylammonium bromide 98%

Supplier: Ambeed
Danger

A119839-100G A119839-25G A119839-500G
77108-508EA 19.57 USD
77108-508 77108-510 77108-512
Tetramethylammonium bromide 98%
Tetramethylammonium bromide

Precaución: for Research Use Only

Formula: C₄H₁₂BrN
MDL Number: MFCD00011626
CAS Number: 64-20-0
UN: 2811
ADR: 6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR