2,4-Dichlorobenzonitrile 96%

Supplier: Ambeed
Danger

A119685-100G A119685-10G A119685-25G
77108-502EA 45.62 USD
77108-502 77108-504 77108-506
2,4-Dichlorobenzonitrile 96%
2,4-Dichlorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃Cl₂N
MW: 172.01 g/mol
MDL Number: MFCD00016373
CAS Number: 6574-98-7
UN: 3439
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR