Fmoc-DL-Phe-OH 97%

Supplier: Ambeed
Warning

A119651-1G A119651-5G
77108-498EA 13.39 USD
77108-498 77108-500
Fmoc-DL-Phe-OH 97%
Fmoc-DL-Phe-OH

Precaución: for Research Use Only

Formula: C₂₄H₂₁NO₄
MW: 387.42 g/mol
MDL Number: MFCD00112977
CAS Number: 126727-04-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR