4-(Aminomethyl)benzonitrile hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A119552-100G
A119552-10G
A119552-1G
A119552-25G
A119552-500G
A119552-5G
77108-478EA
27.81
USD
77108-478
77108-480
77108-482
77108-484
77108-486
77108-488
4-(Aminomethyl)benzonitrile hydrochloride 97%
4-(Aminomethyl)benzonitrile hydrochloride
Precaución: for Research Use Only
Formula:
C₈H₉ClN₂ |
MDL Number:
MFCD01861472 CAS Number: 15996-76-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...