4-Chloro-2-(trifluoromethyl)benzonitrile 96%

Supplier: Ambeed
Danger

Synonyms: 5-Chloro-2-cyanobenzotrifluoride

A119480-1G A119480-5G
77108-474EA 11.33 USD
77108-474 77108-476
4-Chloro-2-(trifluoromethyl)benzonitrile 96%
4-Chloro-2-(trifluoromethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₃ClF₃N
MDL Number: MFCD00060671
CAS Number: 320-41-2
UN: 3276
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR