4,6-Dimethyl-2-mercaptopyrimidine 98% (contains < 15% H₂O)

Supplier: Ambeed
Warning

Synonyms: 2-Mercapto-4,6-dimethylpyrimidine

A119027-25G A119027-5G
77108-452EA 14.86 USD
77108-452 77108-454
4,6-Dimethyl-2-mercaptopyrimidine 98% (contains < 15% H₂O)
4,6-Dimethyl-2-mercaptopyrimidine

Precaución: for Research Use Only

Formula: C₆H₈N₂S
MW: 140.20 g/mol
MDL Number: MFCD00006079
CAS Number: 22325-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR