(5-Chloro-1-methyl-1H-imidazol-2-yl)methanol 95%

Supplier: Ambeed
Warning

Synonyms: 5-Chloro-2-(hydroxymethyl)-1-methyl-1H-imidazole

A118970-100MG A118970-1G A118970-250MG A118970-5G
77108-442EA 53.05 USD
77108-442 77108-444 77108-446 77108-448
(5-Chloro-1-methyl-1H-imidazol-2-yl)methanol 95%
(5-Chloro-1-methyl-1H-imidazol-2-yl)methanol

Precaución: for Research Use Only

Formula: C₅H₇ClN₂O
MW: 146.58 g/mol
MDL Number: MFCD12194768
CAS Number: 334893-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR