1-Bromo-2-(difluoromethoxy)benzene 97%

Supplier: Ambeed
Warning

A118928-100G A118928-25G
77108-434EA 182.48 USD
77108-434 77108-436
1-Bromo-2-(difluoromethoxy)benzene 97%
1-Bromo-2-(difluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₇H₅BrF₂O
MW: 223.02 g/mol
MDL Number: MFCD00221468
CAS Number: 175278-33-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR