2-Amino-2-cyclobutylacetic acid 97%

Supplier: Ambeed
Warning

Synonyms: α-Cyclobutylglycine

A118585-100MG A118585-1G A118585-250MG
77108-424EA 38.2 USD
77108-424 77108-426 77108-428
2-Amino-2-cyclobutylacetic acid 97%
2-Amino-2-cyclobutylacetic acid

Precaución: for Research Use Only

Formula: C₆H₁₁NO₂
MW: 129.157 g/mol
MDL Number: MFCD09264350
CAS Number: 28024-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR