J-147 98%

Supplier: Ambeed
Warning

Synonyms: N-(2,4-dimethylphenyl)-2,2,2-trifluoro-N-[(E)-(3-methoxyphenyl)methylideneamino]acetamide, 2,2,2-Trifluoroacetic acid 1-(2,4-Dimethylphenyl)-2-[(3-methoxyphenyl)methylene]hydrazide, N-(2,4-Dimethylphenyl)-2,2,2-trifluoro-N'-(3-methoxybenzylidene)acetohydrazide

A118455-10MG A118455-1G A118455-250MG A118455-50MG A118455-5G A118455-5MG
77108-412EA 6.37 USD
77108-412 77108-414 77108-416 77108-418 77108-420 77108-422
J-147 98%
J-147

Precaución: for Research Use Only

Formula: C₁₈H₁₇F₃N₂O₂
MW: 350.341 g/mol
MDL Number: MFCD25976644
CAS Number: 1146963-51-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR