tert-Butyl (morpholin-3-ylmethyl)carbamate 95%

Supplier: Ambeed
Warning

A118421-100MG A118421-250MG
77108-408EA 86.53 USD
77108-408 77108-410
tert-Butyl (morpholin-3-ylmethyl)carbamate 95%
tert-Butyl (morpholin-3-ylmethyl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₃
MDL Number: MFCD11111574
CAS Number: 169750-75-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR