2,6-Dichloro-3-fluorobenzoic acid 98%

Supplier: Ambeed
Warning

A117841-100G A117841-1G A117841-25G A117841-5G
77108-354EA 850.79 USD
77108-354 77108-356 77108-358 77108-360
2,6-Dichloro-3-fluorobenzoic acid 98%
2,6-Dichloro-3-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃Cl₂FO₂
MDL Number: MFCD06658165
CAS Number: 178813-78-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR