2-Chloro-5-iodobenzoic acid 98%

Supplier: Ambeed
Warning

A116990-100G A116990-10G A116990-25G A116990-500G A116990-5G
77108-302EA 38.11 USD
77108-302 77108-304 77108-306 77108-308 77108-310
2-Chloro-5-iodobenzoic acid 98%
2-Chloro-5-iodobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄ClIO₂
MDL Number: MFCD00079731
CAS Number: 19094-56-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR