2-Chloro-3,6-difluorobenzaldehyde 98%

Supplier: Ambeed
Warning

A116961-10G A116961-1G A116961-250MG A116961-5G
77108-294EA 43.26 USD
77108-294 77108-296 77108-298 77108-300
2-Chloro-3,6-difluorobenzaldehyde 98%
2-Chloro-3,6-difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃ClF₂O
MDL Number: MFCD01631443
CAS Number: 261762-39-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR