7-Chloro-1,2,3,4-tetrahydroisoquinoline hydrochloride 95%

Supplier: Ambeed
Warning

A116944-100MG A116944-1G A116944-5G
77108-288EA 15.45 USD
77108-288 77108-290 77108-292
7-Chloro-1,2,3,4-tetrahydroisoquinoline hydrochloride 95%
7-Chloro-1,2,3,4-tetrahydroisoquinoline hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₁Cl₂N
MDL Number: MFCD08461076
CAS Number: 73075-45-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR