Z-L-Leu-NH2 97%

Supplier: Ambeed
Warning

Synonyms: (S)-Benzyl (1-amino-4-methyl-1-oxopentan-2-yl)carbamate

A116189-10G A116189-1G A116189-25G A116189-5G
77108-256EA 18.54 USD
77108-256 77108-258 77108-260 77108-262
Z-L-Leu-NH2 97%
Z-L-Leu-NH2

Precaución: for Research Use Only

Formula: C₁₄H₂₀N₂O₃
MW: 264.32 g/mol
MDL Number: MFCD00080252
CAS Number: 4801-79-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR