4-Methoxy-N-(2,4,6-trimethylphenyl)benzenesulfonamide 98%

Supplier: Ambeed
Warning

A115691-10MG A115691-1MG A115691-25MG A115691-5MG A115691-100MG A115691-250MG
77108-248EA 24.73 USD
77108-248 77108-250 77108-252 77108-254 77685-027 77685-028
4-Methoxy-N-(2,4,6-trimethylphenyl)benzenesulfonamide 98%
4-Methoxy-N-(2,4,6-trimethylphenyl)benzenesulfonamide

Precaución: for Research Use Only

Formula: C₁₆H₁₉NO₃S
MDL Number: MFCD02007969
CAS Number: 349085-82-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR