4-Methoxy-3-nitropyridine 98%
Supplier: Ambeed
|
Warning
|
A115235-10G
A115235-1G
A115235-25G
A115235-5G
77108-224EA
32.89
USD
77108-224
77108-226
77108-228
77108-230
4-Methoxy-3-nitropyridine 98%
4-Methoxy-3-nitropyridine
Precaución: for Research Use Only
Formula:
C₆H₆N₂O₃ |
MDL Number:
MFCD00209661 CAS Number: 31872-62-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...