Boc-O-benzyl-D-tyrosine 97%

Supplier: Ambeed
Warning

Synonyms: N-Boc-O-benzyl-D-tyrosine, Boc-D-Tyr(Bzl)-OH

A114252-10G A114252-25G A114252-5G
77108-178EA 70.04 USD
77108-178 77108-180 77108-182
Boc-O-benzyl-D-tyrosine 97%
Boc-O-benzyl-D-tyrosine

Precaución: for Research Use Only

Formula: C₂₁H₂₅NO₅
MW: 371.433 g/mol
MDL Number: MFCD00038249
CAS Number: 63769-58-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR