L(+)-Valine 97%

Supplier: Ambeed
Warning

A114200-100G A114200-25G A114200-500G A114200-1000G
77108-172EA 6.37 USD
77108-172 77108-174 77108-176 77610-918
L(+)-Valine 97%
L(+)-Valine

Precaución: for Research Use Only

Formula: C₅H₁₁NO₂
MDL Number: MFCD00064220
CAS Number: 72-18-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR