N-Methyl-1-(4-(trifluoromethyl)phenyl)methanamine 98%

Supplier: Ambeed
Danger

A114071-1G A114071-250MG A114071-5G
77108-166EA 20.16 USD
77108-166 77108-168 77108-170
N-Methyl-1-(4-(trifluoromethyl)phenyl)methanamine 98%
N-Methyl-1-(4-(trifluoromethyl)phenyl)methanamine

Precaución: for Research Use Only

Formula: C₉H₁₀F₃N
MW: 189.178 g/mol
MDL Number: MFCD04115411
CAS Number: 90390-11-7
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR