3,5-Dibromobenzaldehyde 98%

Supplier: Ambeed
Warning

A113944-100G A113944-10G A113944-1G A113944-25G A113944-500G A113944-5G
77108-154EA 118.45 USD
77108-154 77108-156 77108-158 77108-160 77108-162 77108-164
3,5-Dibromobenzaldehyde 98%
3,5-Dibromobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄Br₂O
MDL Number: MFCD00156887
CAS Number: 56990-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR